Learn More
Analytical Sales & Services SS Tubing 1/16" OD - 0.02" ID x 1M, Yellow
Catalog No. 5000806481
Encompass
Pre-cut Stainless Steel Tubing 1/16" OD - 0.02" ID x 1M, Yellow
Catalog No. 50-008-06481 Supplier Analytical Sales & Services Supplier No. 64135
Pre-cut Stainless Steel Tubing 1/16" OD - 0.02" ID x 1M, Yellow