Learn More
Analytical Sales & Services SS Tubing 1/16" OD - 0.04" ID x 1M
Catalog No. 5000804445
Encompass
Pre-cut Stainless Steel Tubing 1/16" OD - 0.04" ID x 1M
Catalog No. 50-008-04445 Supplier Analytical Sales & Services Supplier No. 64143
Pre-cut Stainless Steel Tubing 1/16" OD - 0.04" ID x 1M