Learn More
Microsolv Technology Corporation Ss Tubing Coil 1/16Th X.010 4
Supplier:  Microsolv Technology Corporation 49410COIL4M
Encompass_Preferred
Tubing, Stainless Steel, Coiled, 0.010 ID, 1/16 OD, HPLC Grade, 4 meters Long. MicroSolv Brand. 1/EA.
Catalog No. 50-014-51685