Learn More
Microsolv Technology Corporation Ss Tubing Coil 1/16Th X.040
Catalog No. 5001451708
Encompass_Preferred
Tubing, Stainless Steel, Coiled, 0.040 ID, 1/16 OD, HPLC Grade, 10 meters Long. MicroSolv Brand. 1/EA.
Catalog No. 50-014-51708 Supplier Microsolv Technology Corporation Supplier No. 49440COIL10M
Tubing, Stainless Steel, Coiled, 0.040 ID, 1/16 OD, HPLC Grade, 10 meters Long. MicroSolv Brand. 1/EA.