Learn More
Waters Corp Waters Tubing Kit, Stainless Steel, 0.030 in. ID, Binary Gradient Module
Catalog No. 50792759 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing kit is designed for the 2525/2545 Binary Gradient Module. It includes all necessary components for installation and operation.
  • Tubing material is stainless steel.
  • Internal diameter of 0.030 in.
  • Includes front-end tubing.
  • Comes with an in-line filter assembly.
  • Screws and brackets are supplied for mounting.
Catalog No. 50-792-759 Supplier Waters Corp Supplier No. 700002491
This tubing kit is designed for the 2525/2545 Binary Gradient Module. It includes all necessary components for installation and operation.
  • Tubing material is stainless steel.
  • Internal diameter of 0.030 in.
  • Includes front-end tubing.
  • Comes with an in-line filter assembly.
  • Screws and brackets are supplied for mounting.