Learn More
Microsolv Technology Corporation Ss Tubing Precut .005X100Cmred
Catalog No. 5001451664
Encompass_Preferred
Tubing, Stainless Steel, Pre-Cut, 0.005 ID, 1/16 OD, HPLC Grade, Red Band, 100cm Long. MicroSolv Brand. 1/EA.
Catalog No. 50-014-51664 Supplier Microsolv Technology Corporation Supplier No. 49405100
Tubing, Stainless Steel, Pre-Cut, 0.005 ID, 1/16 OD, HPLC Grade, Red Band, 100cm Long. MicroSolv Brand. 1/EA.