Learn More
Microsolv Technology Corporation Ss Tubing Precut .005X10Cm Red
Catalog No. 5001451663
Encompass_Preferred
Tubing, Stainless Steel, Pre-Cut, 0.005 ID, 1/16 OD, HPLC Grade, Red Band, 10cm Long. MicroSolv Brand. 1/EA.
Catalog No. 50-014-51663 Supplier Microsolv Technology Corporation Supplier No. 4940510
Tubing, Stainless Steel, Pre-Cut, 0.005 ID, 1/16 OD, HPLC Grade, Red Band, 10cm Long. MicroSolv Brand. 1/EA.