Learn More
Microsolv Technology Corporation Ss Tubing Precut .007X100Cmblk
Catalog No. 5001451672
Encompass_Preferred
Tubing, Stainless Steel, Pre-Cut, 0.007 ID, 1/16 OD, HPLC Grade, Black Band, 100cm Long. MicroSolv Brand. 1/EA.
Catalog No. 50-014-51672 Supplier Microsolv Technology Corporation Supplier No. 49407100
Tubing, Stainless Steel, Pre-Cut, 0.007 ID, 1/16 OD, HPLC Grade, Black Band, 100cm Long. MicroSolv Brand. 1/EA.