Learn More
Microsolv Technology Corporation Ss Tubing Precut .007X50Cm Blk
Catalog No. 5001451675
Encompass_Preferred
Tubing, Stainless Steel, Pre-Cut, 0.007 ID, 1/16 OD, HPLC Grade, Black Band, 50cm Long. MicroSolv Brand. 1/EA.
Catalog No. 50-014-51675 Supplier Microsolv Technology Corporation Supplier No. 4940750
Tubing, Stainless Steel, Pre-Cut, 0.007 ID, 1/16 OD, HPLC Grade, Black Band, 50cm Long. MicroSolv Brand. 1/EA.