Learn More
Microsolv Technology Corporation Ss Tubing Precut .010X5Cm Blue
Catalog No. 5001451678
Encompass_Preferred
Tubing, Stainless Steel, Pre-Cut, 0.010 ID, 1/16 OD, HPLC Grade, Blue Band, 5cm Long. MicroSolv Brand. 1/EA.
Catalog No. 50-014-51678 Supplier Microsolv Technology Corporation Supplier No. 4941005
Tubing, Stainless Steel, Pre-Cut, 0.010 ID, 1/16 OD, HPLC Grade, Blue Band, 5cm Long. MicroSolv Brand. 1/EA.