Learn More
Microsolv Technology Corporation Ss Tubing Precut .020X100Cmyel
Catalog No. 5001451688
Encompass_Preferred
Tubing, Stainless Steel, Pre-Cut, 0.020 ID, 1/16 OD, HPLC Grade, Yellow Band, 100cm Long. MicroSolv Brand. 1/EA.
Catalog No. 50-014-51688 Supplier Microsolv Technology Corporation Supplier No. 49420100
Tubing, Stainless Steel, Pre-Cut, 0.020 ID, 1/16 OD, HPLC Grade, Yellow Band, 100cm Long. MicroSolv Brand. 1/EA.