Learn More
Microsolv Technology Corporation Ss Tubing Precut .030X5Cm Whit
Catalog No. 5001451694
Encompass_Preferred
Tubing, Stainless Steel, Pre-Cut, 0.030 ID, 1/16 OD, HPLC Grade, White Band, 5cm Long. MicroSolv Brand. 1/EA.
Catalog No. 50-014-51694 Supplier Microsolv Technology Corporation Supplier No. 4943005
Tubing, Stainless Steel, Pre-Cut, 0.030 ID, 1/16 OD, HPLC Grade, White Band, 5cm Long. MicroSolv Brand. 1/EA.