Learn More
Microsolv Technology Corporation Ss Tubing Precut .040X20Cmgren
Catalog No. 5001451705
Encompass_Preferred
Tubing, Stainless Steel, Pre-Cut, 0.040 ID, 1/16 OD, HPLC Grade, 20cm Long. MicroSolv Brand. 1/EA.
Catalog No. 50-014-51705 Supplier Microsolv Technology Corporation Supplier No. 4944020
Tubing, Stainless Steel, Pre-Cut, 0.040 ID, 1/16 OD, HPLC Grade, 20cm Long. MicroSolv Brand. 1/EA.