Learn More
Labrepco Inc SS UPRIGHT FRZ DRAW RCK 17.7X8
SDP
Catalog No. NC3240309
Encompass_Preferred
Stainless Steel Upright Freezer Drawer Rack for MATRIX Boxes, FluidX Boxes & Micronic Racks – 15 Box Capacity – 17.7″ (L) x 8″ (H)
Catalog No. NC3240309 Supplier Labrepco Inc Supplier No. UFDMX532
Add to Cart
Add to Cart
Stainless Steel Upright Freezer Drawer Rack for MATRIX Boxes, FluidX Boxes & Micronic Racks – 15 Box Capacity – 17.7″ (L) x 8″ (H)