Learn More
Lakeside Manufacturing Inc SS Utility Cart 3 Shelf: 21x33
Catalog No. 502884508
Encompass_Preferred
Ideal for receiving dock and storage room use, loading and moving laboratory or medical supplies and equipment, and transporting heavy-duty loads up to 1000-Lb. over uneven surfaces, the Lakeside 939 Stainless Steel Utility Cart with (3) shelves measuring 18" x 27" has a weight capacity of 1000-Lb. This Lakeside cart's larger 8” (203) front wheels allow cart to transition over uneven surfaces with ease. Its fixed front wheels help guide cart in a straight line, while making easier to navigate corners. This medical utility cart's angled “u”-shaped frame design provides superior structural strength for larger loads. This utility cart's all joints are full bead welded to better withstand heavy loads and frequent use, while its reinforced edges provide extra capacity for heavy loads. The 939's no-mark, extra load, cushion tread wheels absorb impact, reducing vibration and strain. Bumpers on its legs and handles to protect walls and furniture. NSF listed. Made in the USA.
Catalog No. 50-288-4508 Supplier Lakeside Manufacturing Inc Supplier No. 944
Ideal for receiving dock and storage room use, loading and moving laboratory or medical supplies and equipment, and transporting heavy-duty loads up to 1000-Lb. over uneven surfaces, the Lakeside 939 Stainless Steel Utility Cart with (3) shelves measuring 18" x 27" has a weight capacity of 1000-Lb. This Lakeside cart's larger 8” (203) front wheels allow cart to transition over uneven surfaces with ease. Its fixed front wheels help guide cart in a straight line, while making easier to navigate corners. This medical utility cart's angled “u”-shaped frame design provides superior structural strength for larger loads. This utility cart's all joints are full bead welded to better withstand heavy loads and frequent use, while its reinforced edges provide extra capacity for heavy loads. The 939's no-mark, extra load, cushion tread wheels absorb impact, reducing vibration and strain. Bumpers on its legs and handles to protect walls and furniture. NSF listed. Made in the USA.