Learn More
Thermo Scientific Viper Fingertight Fitting Systems | Stainless Steel | 1 EA
Supplier:  Thermo Scientific 6040.2245
Encompass
Ferrule-free tightening design for virtually zero-dead-volume connections, Operates at pressures up to 1310 bar, Suitable for temperatures up to 120C, Fingertight design, Flexible stainless steel capillaries 1/32 in OD, No tools required, Compatible with valves and columns from virtually any manufacturer, Fits narrow connections such as 10-port valves
Catalog No. NC0996959