Learn More
MCR Safety™ SS1 Black Frame Safety Glasses, Clear UV-AF Lens
Catalog No. 19034357 Shop All MCR Safety Products
Encompass_Preferred
Change view
Click to view available options
Lens Coating:
Anti-fog, Impact-resistant
Catalog No. Lens Coating
19-034-357 Anti-fog, Impact-resistant
1 options
Catalog No. 19-034-357 Supplier MCR Safety™ Supplier No. SS110AF
SS1 Series black safety glasses with clear lens - UV-AF™ anti-fog coating, wrap-around lens design, built-in side shields.
  • Cost-efficient single-lens design
  • Rugged polycarbonate lens filters 99.9% of harmful UV rays
  • Spatula temple with extendable adjustment
  • UV-AF™ Anti-Fog Lens Coating
  • Meets ANSI/ISEA Z87.1 2020 standard
Specifications
Lens Coating Anti-fog, Impact-resistant
Lens Tint Clear
Product Type Glasses
Material Polycarbonate, Nylon
Disposable Reusable
Lens Material Polycarbonate
Frame Material Nylon
UV Protection U6
Frame Type Adjustable Temples
Autoclavable Not Autoclavable