Learn More
Oxidation Technologies LLC SS1 DIFFUSER 1"L X 1/2"D
SDP
Catalog No. NC1908040
Encompass
Sintered Stainless Steel Ozone Diffuser. Bubble ozone into water at flow rates up to 5 LPM. Fine pore bubble diffuser rated at 5 micron. Diffuser area is 1" long x 1/2" in diameter.
Catalog No. NC1908040 Supplier Oxidation Technologies LLC Supplier No. SS1
Sintered Stainless Steel Ozone Diffuser. Bubble ozone into water at flow rates up to 5 LPM. Fine pore bubble diffuser rated at 5 micron. Diffuser area is 1" long x 1/2" in diameter.