Learn More
Grace Bio Labs Inc SS1X13 IMAGING SPACER PK/100
Catalog No. NC0840392
Encompass_Preferred
SS1X13-SecureSeal™ Imaging Spacer, 13mm Diameter ID X 0.12mm Depth, 25mm X 25mm OD - 100 PACK
Catalog No. NC0840392 Supplier Grace Bio Labs Inc Supplier No. 654004
SS1X13-SecureSeal™ Imaging Spacer, 13mm Diameter ID X 0.12mm Depth, 25mm X 25mm OD - 100 PACK