Learn More
Grace Bio Labs Inc SS1X9 IMAGING SPACER PK/100
Catalog No. NC0840391
Encompass_Preferred
SS1X9-SecureSeal™ Imaging Spacer, 9mm Diameter ID X 0.12mm Depth, 18mm X 18mm OD - 100 PACK
Catalog No. NC0840391 Supplier Grace Bio Labs Inc Supplier No. 654002
SS1X9-SecureSeal™ Imaging Spacer, 9mm Diameter ID X 0.12mm Depth, 18mm X 18mm OD - 100 PACK