Learn More
SIMPLEX SCIENCES ss20 DNA Ladder (100µL)
Catalog No. NC2176085
Encompass
The ss20 DNA Ladder is an oligo length standard, or single-stranded DNA ladder, ranging from 10 to 200 nucleotides, with 10-nucleotide increments going from 10 to 100 and 20-nucleotide increments going from 120 to 200.
This item is not returnable. View return policy
Catalog No. NC2176085 Supplier SIMPLEX SCIENCES Supplier No. SS20 DNA LADDER
The ss20 DNA Ladder is an oligo length standard, or single-stranded DNA ladder, ranging from 10 to 200 nucleotides, with 10-nucleotide increments going from 10 to 100 and 20-nucleotide increments going from 120 to 200.