Learn More
Perkin Elmer US LLC SS-316 Tubing - 20 cm x 0.0625 in O.D. x 0.007 in I.D.
Catalog No. 502113473
Encompass_Preferred
Tubing is pre-cut and mirror-polished in 316 stainless steel.
Catalog No. 50-211-3473 Supplier Perkin Elmer US LLC Supplier No. 02711441
Tubing is pre-cut and mirror-polished in 316 stainless steel.