Learn More
LGC GENOMICS LLC (Lucigen) SS320 (MC1061 F') Electrocompetent Cells, 24 Rxns (DUOs)
Supplier: LGC GENOMICS LLC 605122
Construct larger, more complex phage display libraries with highest efficiency cells. SS320 is a non-amber suppressor strain used for phage display.Perfect for antibody phage display or peptide phage display library creation. SS320 Electrocompetent Cells are a non-amber suppressor strain (sometimes called MC1061F') prepared as highly efficient (4 10^10 cfu/g) electrocompetent cells for phage display library screening. These cells have the highest available transformation efficiency of any strain used for phage display.