Learn More
INTACT GENOMICS INC SS320 Phage Display Electrocompetent Cells 6x100µl
Catalog No. 5000082876
Encompass
Intact Genomics SS320 (MC1061F\’) phage display electrocompetent cells are suitable for protein expression, general cloning, blue/white screening, M13 phage work and phage display protein expression.Intact Genomics SS320 phage display electrocompetent cells have the following features: • >5 x 10^10cfu/µg efficiency with electroporation. • Non-amber suppressor strain (sometimes called MC1061F\’)Product Include and Storage: SS320 phage display electrocompetent cells: -80 ºCpUC19 control DNA: -20 ºCRecovery medium: 4ºC
Catalog No. 50-000-82876 Supplier INTACT GENOMICS INC Supplier No. 127424
Intact Genomics SS320 (MC1061F\’) phage display electrocompetent cells are suitable for protein expression, general cloning, blue/white screening, M13 phage work and phage display protein expression.Intact Genomics SS320 phage display electrocompetent cells have the following features: • >5 x 10^10cfu/µg efficiency with electroporation. • Non-amber suppressor strain (sometimes called MC1061F\’)Product Include and Storage: SS320 phage display electrocompetent cells: -80 ºCpUC19 control DNA: -20 ºCRecovery medium: 4ºC