Learn More
INTACT GENOMICS INC SS320 Phage Display Electrocompetent Cells 6x50µl
Supplier:  INTACT GENOMICS INC 117312
Encompass
Intact Genomics SS320 (MC1061F\’) phage display electrocompetent cells are suitable for protein expression, general cloning, blue/white screening, M13 phage work and phage display protein expression.Intact Genomics SS320 phage display electrocompetent cells have the following features: • >5 x 10^10cfu/µg efficiency with electroporation. • Non-amber suppressor strain (sometimes called MC1061F\’)Product Include and Storage: SS320 phage display electrocompetent cells: -80 ºCpUC19 control DNA: -20 ºCRecovery medium: 4ºC
Catalog No. 50-000-82897