Learn More
AMID BIOSCIENCES Amid Biosciences, SS320 Phage Display Chemically Competent E. coli Cells, 10 x 0.1 mL
Supplier: AMID BIOSCIENCES SS32020110
SS320 phage display chemically competent E. coli cells are supplied in a 10 x 0.1 mL format for library preparation and display workflow support. Applications include phage display library propagation, antibody library workflows, peptide display library cloning and rescue. Technical specifications are included below.- Quantity - 10 x 0.1 mL (20 reactions)- Organism - E. coli- Storage - -80 C- Cell Strain / Host - SS320- Cell Format - 10 x 0.1 mL chemically competent cells- Transformation Efficiency - 1.00E+9 This product is for research use only.