Learn More
AMID BIOSCIENCES Amid Biosciences, SS320 Phage Display Electrocompetent E. coli Cells, High Efficiency, 10 x 0.05 mL
Supplier: AMID BIOSCIENCES SS320EC20110
SS320 phage display electrocompetent E. coli cells are supplied in a 10 x 0.05 mL format for high efficiency transformation during library construction and screening workflows. SS320 is commonly used for phage display applications where library size diversity and reproducible recovery are important. Applications include high-efficiency phage display library transformation, antibody library construction, peptide display library workflows. Technical specifications are included below.- Quantity - 10 x 0.05 mL (20 reactions)- Organism - E. coli- Storage - -80 C- Cell Strain / Host - SS320- Cell Format - 10 x 0.05 mL electrocompetent cells- Transformation Efficiency - 1.00E+10This product is for research use only.