Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ss47 Tfa | 00-00-0 | 99.9% | 1067.09 | C51H57F3N6O14S | 5 MG
SDP

Catalog No. 5000222839
Encompass_Preferred

SS47 TFA is a PROTAC-based HPK1 degrader supplied as the trifluoroacetic acid salt for research use. It promotes proteasome-mediated degradation of HPK1 and is used in immuno-oncology and CAR-T cell studies.

  • Research reagent for HPK1 degradation and immuno-oncology studies.
  • Contains an alkyne moiety compatible with copper-catalyzed azide-alkyne cycloaddition.
  • Purity 99.88%.
  • Chemical formula C51H57F3N6O14S; molecular weight 1067.09.
  • Soluble in DMSO at 100 mg/mL; ultrasonic assistance recommended.
  • Store protected from light under nitrogen; in solvent: -80°C (6 months), -20°C (1 month).
  • Available in small research pack sizes such as 1 mg, 5 mg, and 10 mg.

Catalog No. 50-002-22839 Supplier Medchemexpress LLC Supplier No. HY146231A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SS47 TFA is a PROTAC-based HPK1 degrader supplied as the trifluoroacetic acid salt for research use. It promotes proteasome-mediated degradation of HPK1 and is used in immuno-oncology and CAR-T cell studies.

  • Research reagent for HPK1 degradation and immuno-oncology studies.
  • Contains an alkyne moiety compatible with copper-catalyzed azide-alkyne cycloaddition.
  • Purity 99.88%.
  • Chemical formula C51H57F3N6O14S; molecular weight 1067.09.
  • Soluble in DMSO at 100 mg/mL; ultrasonic assistance recommended.
  • Store protected from light under nitrogen; in solvent: -80°C (6 months), -20°C (1 month).
  • Available in small research pack sizes such as 1 mg, 5 mg, and 10 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.