Learn More
SIMPLEX SCIENCES ss50 DNA Ladder (100µL)
Catalog No. NC2175823
Encompass
The ss50 DNA Ladder is an oligo length standard, or single-stranded DNA ladder, ranging from 50 to 500 nucleotides in 50-nucleotide increments.
This item is not returnable. View return policy
Catalog No. NC2175823 Supplier SIMPLEX SCIENCES Supplier No. SS50 DNA LADDER
The ss50 DNA Ladder is an oligo length standard, or single-stranded DNA ladder, ranging from 50 to 500 nucleotides in 50-nucleotide increments.