Learn More
Medchemexpress LLC SSAA09E2 | 883944-52-3 | 98.37% | 300.36 | 50 MG
SDP
Catalog No. 5000348228
Encompass_Preferred
SSAA09E2 is an inhibitor of SARS-CoV (Severe acute respiratory syndrome-Coronavirus) replication. It functions by blocking early interactions between SARS-S and the SARS-CoV receptor, Angiotensin Converting Enzyme-2 (ACE2). This product is intended for research use only.
  • Inhibits SARS-CoV replication
  • Blocks early interactions between SARS-S and ACE2 receptor
  • IC50 (SARS-CoV): 3.1 μM
  • CC50 (HEK-293T): > 100 μM
Catalog No. 50-003-48228 Supplier Medchemexpress LLC Supplier No. HY13806750MG
SSAA09E2 is an inhibitor of SARS-CoV (Severe acute respiratory syndrome-Coronavirus) replication. It functions by blocking early interactions between SARS-S and the SARS-CoV receptor, Angiotensin Converting Enzyme-2 (ACE2). This product is intended for research use only.
  • Inhibits SARS-CoV replication
  • Blocks early interactions between SARS-S and ACE2 receptor
  • IC50 (SARS-CoV): 3.1 μM
  • CC50 (HEK-293T): > 100 μM