Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ssaa09e2 | 883944-52-3 | 98.4% | 300.36 | 100 MG
SDP

Catalog No. 5000352356
Encompass_Preferred

SSAA09E2 is an inhibitor of SARS-CoV (Severe acute respiratory syndrome-Coronavirus) replication. It acts by blocking early interactions of SARS-S with the receptor for SARS-CoV, Angiotensin Converting Enzyme-2 (ACE2).

  • Inhibitor of SARS-CoV replication
  • Blocks early interactions of SARS-S with ACE2 receptor
  • Molecular formula: C16H20N4O2
  • Molecular weight: 300.36
  • Purity: 98.4%
  • Appearance: Solid

Catalog No. 50-003-52356 Supplier Medchemexpress LLC Supplier No. HY138067100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

SSAA09E2 is an inhibitor of SARS-CoV (Severe acute respiratory syndrome-Coronavirus) replication. It acts by blocking early interactions of SARS-S with the receptor for SARS-CoV, Angiotensin Converting Enzyme-2 (ACE2).

  • Inhibitor of SARS-CoV replication
  • Blocks early interactions of SARS-S with ACE2 receptor
  • Molecular formula: C16H20N4O2
  • Molecular weight: 300.36
  • Purity: 98.4%
  • Appearance: Solid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.