Learn More
Medchemexpress LLC Ssaa09e2 | 883944-52-3 | 98.4% | 300.36 g/mol | C16H20N4O2 | 5 MG
SDP
Catalog No. 5000222544
Encompass_Preferred
SSAA09E2 is a small-molecule inhibitor of SARS-CoV replication used in antiviral research. It blocks early interactions between the viral spike (S) protein and the ACE2 receptor, and is supplied as a characterized, high-purity compound suitable for in vitro studies.
  • High purity (98.37%).
  • Well-characterized molecular weight of 300.36 g/mol.
  • Molecular formula C16H20N4O2.
  • Provided in a small quantity suitable for assay development and screening.
  • Intended for in vitro antiviral research applications.
Catalog No. 50-002-22544 Supplier Medchemexpress LLC Supplier No. HY1380675MG
SSAA09E2 is a small-molecule inhibitor of SARS-CoV replication used in antiviral research. It blocks early interactions between the viral spike (S) protein and the ACE2 receptor, and is supplied as a characterized, high-purity compound suitable for in vitro studies.
  • High purity (98.37%).
  • Well-characterized molecular weight of 300.36 g/mol.
  • Molecular formula C16H20N4O2.
  • Provided in a small quantity suitable for assay development and screening.
  • Intended for in vitro antiviral research applications.