Learn More
Next Advance Inc Stainless Steel Beads 1.6 mm RNase Free, 4 mL
SDP
Catalog No. NC1834415
Encompass
RNase free 1.6 mm (1/16 inch) stainless steel beads (7.9 g/cc). In 4 mL package.
Catalog No. NC1834415 Supplier Next Advance Inc Supplier No. SSB16RNA
RNase free 1.6 mm (1/16 inch) stainless steel beads (7.9 g/cc). In 4 mL package.