Learn More
Aviva Systems Biology Corp SSBP1 Antibody - N-terminal region (ARP61972_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61972_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SSBP1. It was validated on Western Blot. . Alternate names: SOSS-B1, SSBP, mtSSB, Mt-SSBApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-287-527