Learn More
Aviva Systems Biology Corp SSBP3 Antibody - middle region (ARP55354_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55354_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SSBP3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SSBP3. Alternate names: CSDP, FLJ10355, SSDP, SSDP1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61476