Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SSC 20x Buffer, MP Biomedicals™

Catalog No. MP11RST2050 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP11RST2050 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP11RST2050 Supplier MP Biomedicals, Inc Supplier No. 11RIST2050
Add to Cart
Add to Cart

SSC 20x Buffer, Molecular biology grade, Clear, Colorless Solution

  • SSC Buffer used in electrophoresis and in sedimentation studies of DNA-histone aggregates.
  • SSC is used as a hybridization buffer in immunoblotting and DNA Microarray protocols.
TRUSTED_SUSTAINABILITY

Specifications

Physical Form Solution
Concentration 20× Concentrated
Color Clear Colorless
Quantity 500 mL
Grade Molecular Biology
storageNote1 Store at Room Temperature (15-30°C).
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.