Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc SSC (20X), 500 mL
SDP

Catalog No. 507517255
Encompass_Preferred

SSC (20X) buffer is a ready-to-use standard reagent in Southern and Northern hybridization procedures Used to control stringency for the washing steps after hybridization. The concentrated formula can be used straight or diluted to make SSC working solutions with concentrations as low as 0.5X.

Applications: SSC (20X) is used for several Cell Biology applications such as: preparing DNA and RNA hybridization buffers and post-hybridization membrane washing buffers, hybridization buffer for Northern and Southern blots, and as a washing buffer for Northern and Southern blots.

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered
DNase, RNase and Protease Tested
Max Shelf Life: 36 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Catalog No. 50-751-7255 Supplier Quality Biological Inc Supplier No. 351003101
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SSC (20X) buffer is a ready-to-use standard reagent in Southern and Northern hybridization procedures Used to control stringency for the washing steps after hybridization. The concentrated formula can be used straight or diluted to make SSC working solutions with concentrations as low as 0.5X.

Applications: SSC (20X) is used for several Cell Biology applications such as: preparing DNA and RNA hybridization buffers and post-hybridization membrane washing buffers, hybridization buffer for Northern and Southern blots, and as a washing buffer for Northern and Southern blots.

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered
DNase, RNase and Protease Tested
Max Shelf Life: 36 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.