Learn More
Biotang Inc SSC (20X, pH 7.0), 2000ml, omposition: Sodium Citrate 0.3M,Sodium Chloride 3.0M,pH 7.0
Catalog No. 507513795 Shop All BIOTANG Inc Products
Encompass_Preferred
SSC (20X, pH 7.0), 2000ml, omposition: Sodium Citrate 0.3M,Sodium Chloride 3.0M,pH 7.0.
Catalog No. 50-751-3795 Supplier Biotang Inc Supplier No. BTBB083
SSC (20X, pH 7.0), 2000ml, omposition: Sodium Citrate 0.3M,Sodium Chloride 3.0M,pH 7.0.