Learn More
Research Products International Corp SSC Buffer 20X Powder, 263.56 Grams of Powder, Makes 1 Liter of 20X Solution
SDP
Catalog No. 50213667
Encompass_Preferred
Dry homogenous blend of molecular biology grade sodium chloride and citric acid, trisodium salt. For use in nucleic acid hybridization at proper working concentration.
Catalog No. 50-213-667 Supplier Research Products International Corp Supplier No. S240201.0
Dry homogenous blend of molecular biology grade sodium chloride and citric acid, trisodium salt. For use in nucleic acid hybridization at proper working concentration.