Learn More
Bioworld SSC Buffer, Ready-Pack (20X), 2 PK(s)
Catalog No. 50196845
Encompass_Preferred
Saline-Sodium Citrate Concentrate Used in protocols for Southern and Northern blots. SSC buffer is used in hybridization cocktails containing DNA and RNA probes for the analyiss of unknown sequences and to locate desired genes and gene fragments. The primary function of SSC is to provide high ionic strength to optimize the rate of probe annealing to the target nucleic acid. The 1X working solution can be easily prepared from the 20X liquid concentrate by diluting with distilled water. Technical Inforamtion:Each pack makes 1 L of 20X- Sodium Citrate concentrateUltra Pure GradeChloride Content (1%, Water)(meq/L): 110-120pH (1%, Water) at 25°C: 7.85-8.15Solubility (1%, Water) (P/F): PassStorage: RTHazard: Irritant
Catalog No. 50-196-845 Supplier Bioworld Supplier No. 107500201
Saline-Sodium Citrate Concentrate Used in protocols for Southern and Northern blots. SSC buffer is used in hybridization cocktails containing DNA and RNA probes for the analyiss of unknown sequences and to locate desired genes and gene fragments. The primary function of SSC is to provide high ionic strength to optimize the rate of probe annealing to the target nucleic acid. The 1X working solution can be easily prepared from the 20X liquid concentrate by diluting with distilled water. Technical Inforamtion:Each pack makes 1 L of 20X- Sodium Citrate concentrateUltra Pure GradeChloride Content (1%, Water)(meq/L): 110-120pH (1%, Water) at 25°C: 7.85-8.15Solubility (1%, Water) (P/F): PassStorage: RTHazard: Irritant