Learn More
Research Products International Corp SSC BUFFER, 20X SOLUTION, 1 LITER
SDP
Catalog No. 50489029
Encompass_Preferred
20X Saline-sodium citrate solution contains:
  • 0.3M sodium citrate
  • 3M NaCl
pH@25°C: 6.85-7.15
Catalog No. 50-489-029 Supplier Research Products International Corp Supplier No. S240221000.0
20X Saline-sodium citrate solution contains:
  • 0.3M sodium citrate
  • 3M NaCl
pH@25°C: 6.85-7.15