Learn More
Crescent Chemical Co Inc SSC BUFFER MOL BIO GRADE
SDP
Catalog No. 50248609
Encompass_Preferred
SSC Buffer molecular biology grade DNase/RNase not detected. 20 x concentrated aqueous solution. Commonly used buffer in transfer, blocking and hybridization in both Northern and Southern Blotting Size - 1L Storage Conditions - +15 °C TO +30 °C /
Catalog No. 50-248-609 Supplier Crescent Chemical Co Inc Supplier No. SE42555.01
SSC Buffer molecular biology grade DNase/RNase not detected. 20 x concentrated aqueous solution. Commonly used buffer in transfer, blocking and hybridization in both Northern and Southern Blotting Size - 1L Storage Conditions - +15 °C TO +30 °C /