Learn More
Bioworld SSC Hybridization Solution (1X), 100 mL
Supplier:  Bioworld 107500221
Encompass_Preferred
SSC Hybridization Solution SSC solution is used for the hybridization of probes to nucleic acids. Contains:6X SSC, 10% Dextran sulfate, 0.2% SDS, 5X Denhardt's solution and sheared salmon sperm DNA
Catalog No. 50-196-846