Learn More
Bioworld SSC Hybridization Solution (1X), 200 mL
Catalog No. 50196847
Encompass_Preferred
SSC Hybridization Solution SSC solution is used for the hybridization of probes to nucleic acids. Contains:6X SSC, 10% Dextran sulfate, 0.2% SDS, 5X Denhardt's solution and sheared salmon sperm DNA
Catalog No. 50-196-847 Supplier Bioworld Supplier No. 107500222
Add to Cart
Add to Cart
SSC Hybridization Solution SSC solution is used for the hybridization of probes to nucleic acids. Contains:6X SSC, 10% Dextran sulfate, 0.2% SDS, 5X Denhardt's solution and sheared salmon sperm DNA