Learn More
Bioworld SSC Prehybridization Solution, 100 mL
Catalog No. 50196849
Encompass_Preferred
SSC Prehybridization Solutions SSC solution is used for the hybridization of probes to nucleic acids. Technical Information:SSC (6X) Denhardt's solution (5X) SDS (0.5%) Denatured, phenol-chloroform extracted from salmon testes DNA (100ug/ml)
Catalog No. 50-196-849 Supplier Bioworld Supplier No. 107500232
SSC Prehybridization Solutions SSC solution is used for the hybridization of probes to nucleic acids. Technical Information:SSC (6X) Denhardt's solution (5X) SDS (0.5%) Denatured, phenol-chloroform extracted from salmon testes DNA (100ug/ml)