Learn More
Medchemexpress LLC SSD114 hydrochloride | 2319790-02-6 | MFCD31619246 | 99.1% | 387.83 | C18H21ClF3N3O | 25 MG
SDP
Catalog No. 5000245171
Encompass_Preferred
SSD114 hydrochloride is a small-molecule positive allosteric modulator of the GABAB receptor intended for in vitro and in vivo research use only.
  • Acts as a GABAB receptor positive allosteric modulator for pharmacological studies.
  • High purity suitable for research applications.
  • Solid, white to off-white appearance for ease of handling.
  • Available in multiple small-scale quantities for experimental flexibility.
  • Stable under recommended storage conditions to preserve compound integrity.
Catalog No. 50-002-45171 Supplier Medchemexpress LLC Supplier No. HY103668A25MG
SSD114 hydrochloride is a small-molecule positive allosteric modulator of the GABAB receptor intended for in vitro and in vivo research use only.
  • Acts as a GABAB receptor positive allosteric modulator for pharmacological studies.
  • High purity suitable for research applications.
  • Solid, white to off-white appearance for ease of handling.
  • Available in multiple small-scale quantities for experimental flexibility.
  • Stable under recommended storage conditions to preserve compound integrity.