Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SSD114 hydrochloride | 2319790-02-6 | MFCD31619246 | 99.1% | 387.83 g/mol | C18H21ClF3N3O | 1 ML
SDP

Catalog No. 5000246811
Encompass_Preferred

SSD114 hydrochloride is a research-grade small molecule described as a novel GABAB receptor positive allosteric modulator for in vitro and preclinical studies.

  • Research use only; not for human therapeutic use.
  • Supplied as a 10 mM solution in DMSO, 1 mL vial.
  • Also available as solid material in multiple weight sizes for formulation or dosing.
  • High purity (≈99.1%), CAS 2319790-02-6, molecular weight 387.83 g/mol.
  • Soluble in DMSO (≈130 mg/mL); limited water solubility.
  • Store sealed, away from moisture; in solvent, keep at -80°C for long-term stability.

Catalog No. 50-002-46811 Supplier Medchemexpress LLC Supplier No. HY103668A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SSD114 hydrochloride is a research-grade small molecule described as a novel GABAB receptor positive allosteric modulator for in vitro and preclinical studies.

  • Research use only; not for human therapeutic use.
  • Supplied as a 10 mM solution in DMSO, 1 mL vial.
  • Also available as solid material in multiple weight sizes for formulation or dosing.
  • High purity (≈99.1%), CAS 2319790-02-6, molecular weight 387.83 g/mol.
  • Soluble in DMSO (≈130 mg/mL); limited water solubility.
  • Store sealed, away from moisture; in solvent, keep at -80°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.