Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp ssDNA Ladder, 10 to 60-mer, Lyophilized, Resolution Check

Catalog No. 502391464 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This ssDNA 10 to 60 Ladder is a lyophilized mixture containing 10 μg of single-stranded DNA components ranging from 10 to 60-mer. It serves as a performance check and potential system suitability standard for LC-UV and LC-MS analyses, supporting various separation modes like IP-RPLC, HILIC, or IEX.

  • Lyophilized mixture of single-stranded DNA.
  • Includes 10, 15, 20, 25, 30, 40, 50, and 60-mer DNA components.
  • Chemically synthesized components confirmed by LC analysis.
  • Performance check and system suitability standard.
  • Compatible with LC-UV and LC-MS analyses.
  • Can be used for IP-RPLC, HILIC, or IEX separations.
  • Store unopened at -20°C ± 10°C.
  • Shipping condition is ambient.
  • Shelf life of 24 months.
  • Reconstituted standard shelf life is 1 week when refrigerated.
  • Requires reconstitution in 100 μL nuclease-free water or buffer.

Catalog No. 50-239-1464 Supplier Waters Corp Supplier No. 186009449
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This ssDNA 10 to 60 Ladder is a lyophilized mixture containing 10 μg of single-stranded DNA components ranging from 10 to 60-mer. It serves as a performance check and potential system suitability standard for LC-UV and LC-MS analyses, supporting various separation modes like IP-RPLC, HILIC, or IEX.

  • Lyophilized mixture of single-stranded DNA.
  • Includes 10, 15, 20, 25, 30, 40, 50, and 60-mer DNA components.
  • Chemically synthesized components confirmed by LC analysis.
  • Performance check and system suitability standard.
  • Compatible with LC-UV and LC-MS analyses.
  • Can be used for IP-RPLC, HILIC, or IEX separations.
  • Store unopened at -20°C ± 10°C.
  • Shipping condition is ambient.
  • Shelf life of 24 months.
  • Reconstituted standard shelf life is 1 week when refrigerated.
  • Requires reconstitution in 100 μL nuclease-free water or buffer.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.