Learn More
AB Sciex LLC ssDNA 100-R Kit
Catalog No. NC1873946
Encompass
For analysis of single-stranded DNA with linearity from 10 to 100 bases. Includes application disk, two coated capillaries, replaceable gel, and standards to perform at least 100 runs. UV detection is required to analyze unlabeled and standard oligonucleotides. Urea denaturant is included separately, enabling a naturing, replaceable gel, if desired. Qty 2: DNA capillary, 65 cm Qty 1: ssDNA 100-R Gel Qty 1: Tris-Borate Buffer Qty 1: 7 M Urea Qty 1: pd(A) 40-60 Test Mix This kit is not compatible with a PDA detector.
Catalog No. NC1873946 Supplier AB Sciex LLC Supplier No. 477480
For analysis of single-stranded DNA with linearity from 10 to 100 bases. Includes application disk, two coated capillaries, replaceable gel, and standards to perform at least 100 runs. UV detection is required to analyze unlabeled and standard oligonucleotides. Urea denaturant is included separately, enabling a naturing, replaceable gel, if desired. Qty 2: DNA capillary, 65 cm Qty 1: ssDNA 100-R Gel Qty 1: Tris-Borate Buffer Qty 1: 7 M Urea Qty 1: pd(A) 40-60 Test Mix This kit is not compatible with a PDA detector.