Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp ssDNA Ladder, 10 μg, 20 to 100-mer, Solid

Catalog No. 502392621 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This product is a lyophilized mixture of single-stranded (ss) DNA molecules ranging from 20 to 100 nucleotides in length. The components are chemically synthesized and verified by LC analysis to be present in reproducible quantities.

  • Quantity: 10 μg
  • Composition: A mixture of 20, 30, 40, 50, 60, 70, 80, 90, and 100-mer single-stranded (ss) DNA.
  • Separation mode: Reversed phase
  • Detection method: MS
  • Formulation: Solid
  • Storage condition: -20°C ±10°C (Freezer)
  • System type: LC/MS
  • Instrument compatibility: UPLC, UHPLC, HPLC
  • Shelf life: 24 months
  • Shelf life upon reconstitution: 1 week
  • Shipping condition: Ambient
  • Tolerance: ±20%
  • Type: Resolution check
  • Application: Oligonucleotide, gene therapeutics
  • Units per package: 1 pk

Catalog No. 50-239-2621 Supplier Waters Corp Supplier No. 186009448
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is a lyophilized mixture of single-stranded (ss) DNA molecules ranging from 20 to 100 nucleotides in length. The components are chemically synthesized and verified by LC analysis to be present in reproducible quantities.

  • Quantity: 10 μg
  • Composition: A mixture of 20, 30, 40, 50, 60, 70, 80, 90, and 100-mer single-stranded (ss) DNA.
  • Separation mode: Reversed phase
  • Detection method: MS
  • Formulation: Solid
  • Storage condition: -20°C ±10°C (Freezer)
  • System type: LC/MS
  • Instrument compatibility: UPLC, UHPLC, HPLC
  • Shelf life: 24 months
  • Shelf life upon reconstitution: 1 week
  • Shipping condition: Ambient
  • Tolerance: ±20%
  • Type: Resolution check
  • Application: Oligonucleotide, gene therapeutics
  • Units per package: 1 pk

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.